Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017778.1 | NP_001017778.1 | 2 - 3 | 209 | 71 | |
GenBank mRNA | BC092901.1 | - | 2 - 3 | 209 | 71 | |
BC164579.1 | - | 1 - 2 | 165 | 71 | ||
HM239643.1 | - | 1 - 2 | 53 | 71 | ||
HM239644.1 | - | 1 - 2 | 53 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |