Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017846.1 | NP_001017846.1 | 3 - 4 | 259 | 94 | |
GenBank mRNA | BC092767.1 | - | 3 - 4 | 259 | 94 | |
BC164299.1 | - | 3 - 4 | 254 | 94 | ||
BC171682.1 | - | 3 - 4 | 245 | 94 | ||
BC171686.1 | - | 3 - 4 | 245 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |