Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017871.1 | NP_001017871.1 | 2 - 3 | 147 | 94 | |
GenBank mRNA | BC092977.1 | - | 2 - 3 | 147 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |