Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018126.2 | NP_001018136.2 | 4 - 5 | 945 | 88 | |
XM_005163053.3 | XP_005163110.1 | 4 - 5 | 889 | 88 | ||
XM_005163054.3 | XP_005163111.1 | 4 - 5 | 883 | 88 | ||
GenBank mRNA | BC095641.1 | - | 4 - 5 | 951 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |