Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tma16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177745.1 | NP_001171216.1 | 6 - 7 | 535 | 91 | |
GenBank mRNA | BC095357.1 | - | 6 - 7 | 534 | 91 | |
BC152117.1 | - | 6 - 7 | 494 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |