Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stxbp6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020738.2 | NP_001018574.2 | 3 - 4 | 419 | 66 | |
XM_005160403.4 | XP_005160460.1 | 5 - 6 | 904 | 66 | ||
XM_009294576.3 | XP_009292851.1 | 5 - 6 | 905 | 66 | ||
XM_021468334.1 | XP_021324009.1 | 4 - 5 | 773 | 66 | ||
GenBank mRNA | BC095177.1 | - | 3 - 4 | 419 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |