Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024373.1 | NP_001019544.1 | 3 - 4 | 371 | 86 | |
GenBank mRNA | BC095392.1 | - | 3 - 4 | 371 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |