Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp5f1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024429.2 | NP_001019600.2 | 8 - 9 | 1336 | 77 | |
GenBank mRNA | BC095620.1 | - | 8 - 9 | 77 | ||
BC154659.1 | - | 8 - 9 | 77 | |||
EH510787.1 | - | 2 - 3 | 466 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |