Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif1axb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025453.1 | NP_001020624.1 | 3 - 4 | 346 | 84 | |
GenBank mRNA | BC096794.1 | - | 3 - 4 | 346 | 84 | |
BC155227.1 | - | 3 - 4 | 343 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |