Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025470.1 | NP_001020641.1 | 2 - 3 | 364 | 74 | |
GenBank mRNA | BC097166.1 | - | 2 - 3 | 364 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |