Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025470.1 | NP_001020641.1 | 3 - 4 | 934 | 111 | |
GenBank mRNA | BC097166.1 | - | 3 - 4 | 934 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |