Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brms1la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030102.2 | NP_001025273.1 | 7 - 8 | 1055 | 117 | |
XM_005158647.4 | XP_005158704.1 | 7 - 8 | 1033 | 131 | ||
GenBank mRNA | BC097187.1 | - | 7 - 8 | 1063 | 117 | |
EB900863.1 | - | 1 - 2 | 63 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |