Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: camk2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030125.1 | NP_001025296.1 | 20 - 21 | 1777 | 78 | |
XM_005156361.4 | XP_005156418.1 | 22 - 23 | 2025 | 78 | ||
XM_005156362.4 | XP_005156419.1 | 22 - 23 | 2016 | 78 | ||
XM_005156363.4 | XP_005156420.1 | 21 - 22 | 1962 | 78 | ||
XM_005156364.4 | XP_005156421.1 | 21 - 22 | 1939 | 78 | ||
XM_005156365.4 | XP_005156422.1 | 21 - 22 | 1886 | 78 | ||
XM_005156366.4 | XP_005156423.1 | 20 - 21 | 1866 | 78 | ||
XM_005156367.4 | XP_005156424.1 | 20 - 21 | 1781 | 78 | ||
XM_005156368.4 | XP_005156425.1 | 20 - 21 | 1771 | 78 | ||
XM_005156369.4 | XP_005156426.1 | 19 - 20 | 1708 | 78 | ||
XM_005156370.4 | XP_005156427.1 | 19 - 20 | 1685 | 78 | ||
XM_005156371.4 | XP_005156428.1 | 18 - 19 | 1621 | 78 | ||
XM_009306817.3 | XP_009305092.1 | 20 - 21 | 1875 | 78 | ||
XM_021480164.1 | XP_021335839.1 | 21 - 22 | 1929 | 78 | ||
XM_021480165.1 | XP_021335840.1 | 21 - 22 | 1876 | 78 | ||
XM_021480166.1 | XP_021335841.1 | 19 - 20 | 1694 | 78 | ||
GenBank mRNA | BC096785.1 | - | 20 - 21 | 1777 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |