Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: camk2g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030125.1 | NP_001025296.1 | 14 - 15 | 1150 | 87 | |
XM_005156361.4 | XP_005156418.1 | 15 - 16 | 1260 | 87 | ||
XM_005156362.4 | XP_005156419.1 | 15 - 16 | 1260 | 87 | ||
XM_005156363.4 | XP_005156420.1 | 14 - 15 | 1197 | 87 | ||
XM_005156365.4 | XP_005156422.1 | 15 - 16 | 1259 | 87 | ||
XM_005156367.4 | XP_005156424.1 | 15 - 16 | 1259 | 87 | ||
XM_005156368.4 | XP_005156425.1 | 15 - 16 | 1258 | 87 | ||
XM_005156369.4 | XP_005156426.1 | 14 - 15 | 1195 | 87 | ||
XM_021480165.1 | XP_021335840.1 | 15 - 16 | 1258 | 87 | ||
GenBank mRNA | BC096785.1 | - | 14 - 15 | 1150 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |