Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320148.1 | NP_001307077.1 | 14 - 15 | 2262 | 102 | |
XM_017354091.2 | XP_017209580.1 | 14 - 15 | 2305 | 102 | ||
GenBank mRNA | BC146747.1 | - | 14 - 15 | 2258 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |