Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem51b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102617.1 | NP_001096087.1 | 2 - 3 | 645 | 77 | |
GenBank mRNA | BC150370.1 | - | 2 - 3 | 645 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |