Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: capns1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109706.1 | NP_001103176.1 | 4 - 5 | 358 | 143 | |
XM_005157735.1 | XP_005157792.1 | 4 - 5 | 298 | 143 | ||
GenBank mRNA | BC151952.1 | - | 4 - 5 | 358 | 143 | |
BC162123.1 | - | 4 - 5 | 344 | 143 | ||
BC162479.1 | - | 4 - 5 | 344 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |