Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002041.1 | NP_001002041.1 | 14 - 15 | 1440 | 61 | |
GenBank mRNA | AY648765.1 | - | 14 - 15 | 1440 | 61 | |
BC045326.1 | - | 14 - 15 | 1441 | 61 | ||
BC163908.1 | - | 14 - 15 | 1429 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |