Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002041.1 | NP_001002041.1 | 9 - 10 | 841 | 96 | |
GenBank mRNA | AY648765.1 | - | 9 - 10 | 841 | 96 | |
BC045326.1 | - | 9 - 10 | 842 | 96 | ||
BC163908.1 | - | 9 - 10 | 830 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |