Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2aa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002092.2 | NP_001002092.1 | 3 - 4 | 531 | 93 | |
XM_017353539.2 | XP_017209028.1 | 3 - 4 | 524 | 93 | ||
GenBank mRNA | AB303423.1 | - | 3 - 4 | 528 | 93 | |
BC071533.1 | - | 3 - 4 | 533 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |