Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105599.1 | NP_001099069.1 | 4 - 5 | 423 | 134 | |
XM_021479212.1 | XP_021334887.1 | 4 - 5 | 473 | 134 | ||
GenBank mRNA | BC071425.1 | - | 4 - 5 | 422 | 134 | |
BC153595.1 | - | 4 - 5 | 423 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |