Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002139.1 | NP_001002139.1 | 3 - 4 | 283 | 77 | |
GenBank mRNA | AY648841.1 | - | 3 - 4 | 286 | 77 | |
BC071420.1 | - | 3 - 4 | 283 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |