Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002343.1 | NP_001002343.1 | 6 - 7 | 1054 | 101 | |
XM_005171375.4 | XP_005171432.1 | 6 - 7 | 1228 | 101 | ||
XM_021479745.1 | XP_021335420.1 | 6 - 7 | 1224 | 101 | ||
GenBank mRNA | BC075920.1 | - | 6 - 7 | 1054 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |