Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tm4sf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002372.1 | NP_001002372.1 | 4 - 5 | 673 | 132 | |
GenBank mRNA | BC075884.1 | - | 4 - 5 | 673 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |