Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crygm2d13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002408.1 | NP_001002408.1 | 2 - 3 | 301 | 120 | |
GenBank mRNA | BC076172.1 | - | 2 - 3 | 301 | 120 | |
BC152692.1 | - | 2 - 3 | 288 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |