Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002436.2 | NP_001002436.2 | 5 - 6 | 800 | 102 | |
XM_005169066.4 | XP_005169123.1 | 5 - 6 | 786 | 102 | ||
GenBank mRNA | AI437369.1 | - | 1 - 2 | 153 | 102 | |
BC076129.1 | - | 5 - 6 | 766 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |