Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002447.1 | NP_001002447.1 | 5 - 6 | 573 | 94 | |
XM_005156283.4 | XP_005156340.1 | 5 - 6 | 691 | 94 | ||
XM_005156285.3 | XP_005156342.1 | 5 - 6 | 699 | 94 | ||
XM_017358446.2 | XP_017213935.1 | 5 - 6 | 615 | 94 | ||
GenBank mRNA | BC076024.1 | - | 5 - 6 | 573 | 94 | |
BC152175.1 | - | 5 - 6 | 669 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |