Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gatd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037118.1 | NP_001032195.1 | 2 - 3 | 321 | 94 | |
GenBank mRNA | BC085644.1 | - | 2 - 3 | 150 | 94 | |
BC107609.1 | - | 2 - 3 | 321 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |