Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037699.1 | NP_001032788.1 | 7 - 8 | 2233 | 77 | |
GenBank mRNA | BC107999.1 | - | 7 - 8 | 2233 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |