Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc8a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039144.1 | NP_001034233.1 | 3 - 4 | 2081 | 91 | |
XM_005158696.4 | XP_005158753.1 | 4 - 5 | 2631 | 91 | ||
XM_005158697.4 | XP_005158754.1 | 4 - 5 | 2623 | 91 | ||
XM_009293025.3 | XP_009291300.1 | 4 - 5 | 2629 | 91 | ||
XM_021467243.1 | XP_021322918.1 | 2 - 3 | 313 | 91 | ||
GenBank mRNA | AY934776.1 | - | 3 - 4 | 2081 | 91 | |
BC163419.1 | - | 3 - 4 | 2108 | 91 | ||
EU559286.1 | - | 4 - 5 | 2274 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |