Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126411.1 | NP_001119883.1 | 3 - 4 | 631 | 103 | |
XM_005167854.4 | XP_005167911.1 | 3 - 4 | 657 | 103 | ||
GenBank mRNA | BC107618.1 | - | 3 - 4 | 642 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |