Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042694.2 | NP_001036159.2 | 9 - 10 | 1333 | 58 | |
GenBank mRNA | BC134110.1 | - | 9 - 10 | 1333 | 58 | |
DQ826745.1 | - | 8 - 9 | 1221 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |