Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: top1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044324.2 | NP_001037789.1 | 6 - 7 | 528 | 100 | |
GenBank mRNA | BC071418.1 | - | 6 - 7 | 527 | 100 | |
BC095049.1 | - | 6 - 7 | 487 | 100 | ||
BC117588.1 | - | 6 - 7 | 528 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |