Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044811.2 | NP_001038276.1 | 4 - 5 | 672 | 83 | |
GenBank mRNA | BC133967.1 | - | 4 - 5 | 672 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |