Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044816.1 | NP_001038281.1 | 4 - 5 | 488 | 71 | |
XM_009306499.2 | XP_009304774.1 | 6 - 7 | 1293 | 71 | ||
XM_009306500.2 | XP_009304775.1 | 5 - 6 | 1168 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |