Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045374.2 | NP_001038839.2 | 1 - 2 | 178 | 76 | |
GenBank mRNA | BC122395.1 | - | 1 - 2 | 147 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |