Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045374.2 | NP_001038839.2 | 5 - 6 | 570 | 118 | |
GenBank mRNA | BC122395.1 | - | 5 - 6 | 539 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |