Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045854.2 | NP_001039319.1 | 2 - 3 | 376 | 86 | |
GenBank mRNA | BC122270.1 | - | 2 - 3 | 377 | 86 | |
BC164608.1 | - | 2 - 3 | 207 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |