Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oxct1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077150.1 | NP_001070618.1 | 7 - 8 | 807 | 91 | |
GenBank mRNA | BC124298.1 | - | 7 - 8 | 807 | 91 | |
BC154254.1 | - | 7 - 8 | 789 | 91 | ||
BC155216.1 | - | 7 - 8 | 829 | 91 | ||
BC164688.1 | - | 7 - 8 | 791 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |