Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kctd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077151.1 | NP_001070619.1 | 5 - 6 | 743 | 97 | |
GenBank mRNA | BC122284.1 | - | 5 - 6 | 743 | 97 | |
BC157374.1 | - | 5 - 6 | 711 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |