Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr02 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077158.2 | NP_001070626.2 | 2 - 3 | 360 | 97 | |
GenBank mRNA | BC122154.1 | - | 2 - 3 | 97 | ||
BC152089.1 | - | 2 - 3 | 360 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |