Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmga1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077276.1 | NP_001070744.1 | 2 - 3 | 254 | 115 | |
XM_017356518.2 | XP_017212007.1 | 2 - 3 | 646 | 115 | ||
GenBank mRNA | BC124777.1 | - | 2 - 3 | 289 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |