Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077295.2 | NP_001070763.1 | 4 - 5 | 362 | 103 | |
GenBank mRNA | AW420067.1 | - | 4 - 5 | 321 | 103 | |
BC124718.1 | - | 4 - 5 | 363 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |