Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2x9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077453.1 | NP_001070921.1 | 4 - 5 | 673 | 126 | |
GenBank mRNA | BC124413.1 | - | 4 - 5 | 673 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |