Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noxo1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077584.1 | NP_001071052.1 | 4 - 5 | 481 | 79 | |
GenBank mRNA | AI959305.1 | - | 4 - 5 | 461 | 79 | |
BC124172.1 | - | 4 - 5 | 481 | 79 | ||
BI326762.1 | - | 3 - 4 | 330 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |