Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077615.1 | NP_001071083.1 | 6 - 7 | 1555 | 62 | |
XM_005158452.4 | XP_005158509.1 | 6 - 7 | 1782 | 62 | ||
XM_005158453.4 | XP_005158510.1 | 6 - 7 | 1794 | 62 | ||
GenBank mRNA | AB361066.1 | - | 5 - 6 | 1386 | 62 | |
BC124788.1 | - | 6 - 7 | 1555 | 62 | ||
BC155730.1 | - | 6 - 7 | 1567 | 62 | ||
BC165400.1 | - | 5 - 6 | 1420 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |