Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077770.1 | NP_001071238.1 | 315 | 143 | ||
GenBank mRNA | BC095388.1 | - | 315 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |