Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmgcra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079977.2 | NP_001073446.2 | 15 - 16 | 2194 | 74 | |
XM_005165515.4 | XP_005165572.1 | 15 - 16 | 2169 | 74 | ||
GenBank mRNA | BC117622.1 | - | 15 - 16 | 2198 | 74 | |
BC155135.1 | - | 15 - 16 | 2169 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |