Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080081.1 | NP_001073550.1 | 5 - 6 | 650 | 96 | |
XM_009297718.3 | XP_009295993.1 | 5 - 6 | 96 | |||
GenBank mRNA | BC128877.1 | - | 5 - 6 | 650 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |