Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp3ccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080594.2 | NP_001074063.2 | 11 - 12 | 1530 | 114 | |
XM_005155313.4 | XP_005155370.1 | 11 - 12 | 1576 | 114 | ||
XM_005155314.4 | XP_005155371.1 | 11 - 12 | 1576 | 114 | ||
GenBank mRNA | BC129382.1 | - | 11 - 12 | 1521 | 114 | |
BC165627.1 | - | 11 - 12 | 1251 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |